1-Bromo-5-chloro-4-ethoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

54157-500MG 54157-1G 54157-5G 54157-25G
77329-204EA 275.01 USD
77329-204 77353-018 77365-492 77387-478
1-Bromo-5-chloro-4-ethoxy-2-methylbenzene ≥95%
1-Bromo-5-chloro-4-ethoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21993047
CAS Number: 1368775-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR