1-Chloro-5-iodo-2-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

54156-500MG 54156-1G 54156-5G 54156-25G
77329-202EA 155.53 USD
77329-202 77345-564 77365-490 77387-476
1-Chloro-5-iodo-2-methoxy-4-methylbenzene ≥97%
1-Chloro-5-iodo-2-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
MW: 282.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32704830
CAS Number: 2383588-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR