(5-Chloro-4-methoxy-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54155-500MG 54155-1G 54155-5G 54155-25G
77329-200EA 194.67 USD
77329-200 77353-016 77365-488 77387-474
(5-Chloro-4-methoxy-2-methylphenyl)(methyl)sulfane ≥97%
(5-Chloro-4-methoxy-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClOS
Storage Temperature: Ambient
MDL Number: MFCD34564252
CAS Number: 2707609-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR