5-Chloro-4-methoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54153-500MG 54153-1G 54153-5G 54153-25G
77329-196EA 92.7 USD
77329-196 77342-466 77365-484 77377-258
5-Chloro-4-methoxy-2-methylbenzaldehyde ≥97%
5-Chloro-4-methoxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246305
CAS Number: 883529-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR