Ethyl 5-chloro-4-methoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

54152-500MG 54152-1G 54152-5G
77335-474EA 355.35 USD
77335-474 77353-012 77377-256
Ethyl 5-chloro-4-methoxy-2-methylbenzoate ≥97%
Ethyl 5-chloro-4-methoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34621881
CAS Number: 1417735-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR