2,5-Dichloro-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54130-500MG 54130-1G 54130-5G
77338-416EA 487.19 USD
77338-416 77353-006 77377-254
2,5-Dichloro-3-(methylthio)benzaldehyde ≥97%
2,5-Dichloro-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂OS
MW: 221.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564251
CAS Number: 2734776-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR