2,5-Dichloro-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

54127-500MG 54127-1G 54127-25G 54127-5G
77329-190EA 295.61 USD
77329-190 77353-004 77387-466 77377-252
2,5-Dichloro-3-(methylthio)benzoic acid ≥97%
2,5-Dichloro-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34757237
CAS Number: 2764728-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR