2-Fluoro-5-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

54124-500MG 54124-1G 54124-5G 54124-25G
77329-188EA 276.04 USD
77329-188 77353-002 77365-478 77387-464
2-Fluoro-5-methyl-1,1'-biphenyl ≥97%
2-Fluoro-5-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁F
MW: 186.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701864
CAS Number: 1214342-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR