2-Fluoro-3-iodo-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54114-500MG 54114-1G 54114-5G 54114-25G
77329-182EA 122.57 USD
77329-182 77343-932 77365-472 77377-246
2-Fluoro-3-iodo-5-methylbenzoic acid ≥97%
2-Fluoro-3-iodo-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757226
CAS Number: 2385438-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR