Ethyl 2-fluoro-3,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

54101-500MG 54101-1G 54101-5G
77338-146EA 466.59 USD
77338-146 77352-996 77377-238
Ethyl 2-fluoro-3,5-dimethylbenzoate ≥97%
Ethyl 2-fluoro-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
MW: 196.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794290
CAS Number: 1806332-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR