4,4,5,5-Tetramethyl-2-(2,4,5-trimethoxyphenyl)-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

54097-500MG 54097-1G 54097-5G
77329-174EA 326.51 USD
77329-174 77352-994 77377-236
4,4,5,5-Tetramethyl-2-(2,4,5-trimethoxyphenyl)-1,3,2-dioxaborolane ≥97%
4,4,5,5-Tetramethyl-2-(2,4,5-trimethoxyphenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₅
MW: 294.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757214
CAS Number: 2048455-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR