Methyl(2,4,5-trimethoxyphenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54093-500MG 54093-1G 54093-5G 54093-25G
77329-172EA 157.59 USD
77329-172 77345-700 77365-464 77387-458
Methyl(2,4,5-trimethoxyphenyl)sulfane ≥97%
Methyl(2,4,5-trimethoxyphenyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃S
MW: 214.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757213
CAS Number: 2570-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR