1-Iodo-2,4,5-trimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

54092-500MG 54092-1G 54092-5G 54092-25G
77329-170EA 146.26 USD
77329-170 77345-168 77365-462 77387-456
1-Iodo-2,4,5-trimethoxybenzene ≥97%
1-Iodo-2,4,5-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO₃
MW: 294.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835332
CAS Number: 23149-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR