1-Iodo-3-methoxy-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

53991-500MG 53991-1G 53991-5G 53991-25G
77329-164EA 243.08 USD
77329-164 77352-988 77365-458 77387-452
1-Iodo-3-methoxy-5-methylbenzene ≥95%
1-Iodo-3-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₉IO
Storage Temperature: Ambient
MDL Number: MFCD18397769
CAS Number: 1261826-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR