4-Ethoxy-3-fluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53986-500MG 53986-1G 53986-5G
77339-032EA 545.9 USD
77339-032 77352-986 77377-232
4-Ethoxy-3-fluoro-5-methoxybenzoic acid ≥97%
4-Ethoxy-3-fluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
MW: 214.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564249
CAS Number: 2385432-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR