3-Chloro-2-ethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

53979-1G 53979-5G 53979-25G
77341-342EA 82.4 USD
77341-342 77365-456 77377-230
3-Chloro-2-ethoxy-1,1'-biphenyl ≥97%
3-Chloro-2-ethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO
MW: 232.705 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757206
CAS Number: 2764728-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR