2-(Benzyloxy)-3-chloro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

53978-500MG 53978-1G 53978-5G 53978-25G
77329-162EA 144.2 USD
77329-162 77345-018 77365-454 77387-450
2-(Benzyloxy)-3-chloro-1,1'-biphenyl ≥97%
2-(Benzyloxy)-3-chloro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₉H₁₅ClO
MW: 294.775 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757205
CAS Number: 855751-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR