5-Chloro-6-methoxy-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

53973-500MG 53973-1G 53973-5G
77329-160EA 324.45 USD
77329-160 77352-984 77377-228
5-Chloro-6-methoxy-[1,1'-biphenyl]-3-carbaldehyde ≥97%
5-Chloro-6-methoxy-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
MW: 246.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757200
CAS Number: 2901082-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR