5-Chloro-6-methoxy-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

53970-500MG 53970-1G 53970-25G 53970-5G
77329-158EA 302.82 USD
77329-158 77352-982 77387-448 77377-226
5-Chloro-6-methoxy-[1,1'-biphenyl]-3-carboxylic acid ≥97%
5-Chloro-6-methoxy-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₃
MW: 262.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757197
CAS Number: 107517-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR