5-Bromo-3-chloro-2-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

53967-500MG 53967-1G 53967-5G 53967-25G
77329-154EA 90.64 USD
77329-154 77342-392 77365-450 77377-224
5-Bromo-3-chloro-2-methoxy-1,1'-biphenyl ≥97%
5-Bromo-3-chloro-2-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrClO
MW: 297.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757194
CAS Number: 2807445-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR