5-Bromo-3-chloro-[1,1'-biphenyl]-2-ol ≥97%

Supplier: AOBChem USA

Warning

53966-1G 53966-5G 53966-25G
77341-028EA 66.95 USD
77341-028 77365-448 77377-222
5-Bromo-3-chloro-[1,1'-biphenyl]-2-ol ≥97%
5-Bromo-3-chloro-[1,1'-biphenyl]-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₈BrClO
MW: 283.548 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757193
CAS Number: 2764730-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR