(3-Chloro-2,4-difluorophenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

53965-1G 53965-5G 53965-25G
77340-730EA 60.77 USD
77340-730 77365-446 77377-220
(3-Chloro-2,4-difluorophenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(3-Chloro-2,4-difluorophenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClF₂N₂O
MW: 274.694 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757192
CAS Number: 2764730-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR