(3-Chloro-2,4-difluorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

53964-1G 53964-5G 53964-25G
77340-728EA 60.77 USD
77340-728 77365-444 77377-218
(3-Chloro-2,4-difluorophenyl)(thiomorpholino)methanone ≥97%
(3-Chloro-2,4-difluorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClF₂NOS
MW: 277.718 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757191
CAS Number: 2764733-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR