(3-Chloro-2,4-difluorophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

53963-1G 53963-5G 53963-25G
77340-726EA 60.77 USD
77340-726 77365-442 77377-216
(3-Chloro-2,4-difluorophenyl)(morpholino)methanone ≥97%
(3-Chloro-2,4-difluorophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClF₂NO₂
MW: 261.652 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757190
CAS Number: 2764732-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR