3-Chloro-2,4-difluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

53957-1G 53957-5G 53957-25G
77341-026EA 66.95 USD
77341-026 77365-432 77377-206
3-Chloro-2,4-difluoro-N-methylbenzamide ≥97%
3-Chloro-2,4-difluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₆ClF₂NO
Storage Temperature: Ambient
MDL Number: MFCD30161674
CAS Number: 1864925-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR