(5-Chloro-2-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53951-500MG 53951-1G 53951-5G
77329-152EA 318.27 USD
77329-152 77352-976 77377-202
(5-Chloro-2-isopropoxyphenyl)methanol ≥97%
(5-Chloro-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO₂
MW: 200.662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11180662
CAS Number: 23426-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR