2-Chloro-3-fluoro-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

53946-500MG 53946-1G 53946-5G
77338-252EA 470.71 USD
77338-252 77352-972 77377-198
2-Chloro-3-fluoro-4-phenylpyridine ≥97%
2-Chloro-3-fluoro-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₇ClFN
MW: 207.631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752347
CAS Number: 1805522-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR