(E)-5-(trifluoromethyl)picolinaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

53943-500MG 53943-1G 53943-5G 53943-25G
77329-148EA 145.23 USD
77329-148 77345-080 77365-430 77387-442
(E)-5-(trifluoromethyl)picolinaldehyde oxime ≥97%
(E)-5-(trifluoromethyl)picolinaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O
MW: 190.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20277974
CAS Number: 1280538-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR