1-Chloro-2-methoxy-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

53942-500MG 53942-1G 53942-5G
77336-744EA 393.46 USD
77336-744 77352-970 77377-196
1-Chloro-2-methoxy-3,5-dimethylbenzene ≥97%
1-Chloro-2-methoxy-3,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
MW: 170.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751476
CAS Number: 1266515-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR