1-Chloro-2-isopropoxy-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

53940-500MG 53940-1G 53940-5G
77338-840EA 524.27 USD
77338-840 77352-966 77377-192
1-Chloro-2-isopropoxy-3,5-dimethylbenzene ≥97%
1-Chloro-2-isopropoxy-3,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClO
MW: 198.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530240
CAS Number: 1266457-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR