1-Bromo-2-(methoxymethoxy)-3,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

53936-1G 53936-5G 53936-25G
77341-930EA 108.15 USD
77341-930 77365-428 77377-186
1-Bromo-2-(methoxymethoxy)-3,5-dimethylbenzene ≥95%
1-Bromo-2-(methoxymethoxy)-3,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757184
CAS Number: 2589880-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR