2-(Benzyloxy)-3,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53934-500MG 53934-1G 53934-5G
77336-874EA 396.55 USD
77336-874 77352-958 77377-182
2-(Benzyloxy)-3,5-dimethylbenzaldehyde ≥97%
2-(Benzyloxy)-3,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD30297287
CAS Number: 1386379-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR