Methyl 2-(benzyloxy)-3,5-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

53932-500MG 53932-1G 53932-5G
77329-144EA 338.87 USD
77329-144 77352-954 77377-178
Methyl 2-(benzyloxy)-3,5-dimethylbenzoate ≥95%
Methyl 2-(benzyloxy)-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757181
CAS Number: 2755718-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR