2-(Benzyloxy)-1-bromo-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

53927-500MG 53927-1G 53927-5G 53927-25G
77329-138EA 147.29 USD
77329-138 77345-212 77365-426 77387-438
2-(Benzyloxy)-1-bromo-3,5-dimethylbenzene ≥97%
2-(Benzyloxy)-1-bromo-3,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
MW: 291.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530236
CAS Number: 2734772-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR