2-Isopropoxy-3,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53925-500MG 53925-1G 53925-5G
77336-872EA 396.55 USD
77336-872 77352-948 77377-172
2-Isopropoxy-3,5-dimethylbenzaldehyde ≥97%
2-Isopropoxy-3,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD31504561
CAS Number: 1159697-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR