Ethyl 2-ethoxy-3,5-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

53918-500MG 53918-1G 53918-25G 53918-5G
77329-136EA 300.76 USD
77329-136 77352-944 77387-436 77377-168
Ethyl 2-ethoxy-3,5-dimethylbenzoate ≥95%
Ethyl 2-ethoxy-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31773420
CAS Number: 1369368-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR