Methyl 2-ethoxy-3,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

53917-500MG 53917-1G 53917-25G 53917-5G
77329-134EA 300.76 USD
77329-134 77352-942 77387-434 77377-166
Methyl 2-ethoxy-3,5-dimethylbenzoate ≥97%
Methyl 2-ethoxy-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD31773419
CAS Number: 1368199-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR