(2-Ethoxy-3,5-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

53912-500MG 53912-1G 53912-5G
77337-668EA 435.69 USD
77337-668 77352-940 77377-164
(2-Ethoxy-3,5-dimethylphenyl)boronic acid ≥97%
(2-Ethoxy-3,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381032
CAS Number: 1334221-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR