2-(2-Methoxy-3,5-dimethylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

53911-500MG 53911-1G 53911-5G
77340-310EA 900.22 USD
77340-310 77352-938 77377-162
2-(2-Methoxy-3,5-dimethylphenyl)propan-2-ol ≥97%
2-(2-Methoxy-3,5-dimethylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757176
CAS Number: 79416-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR