1-(2-Methoxy-3,5-dimethylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

53909-500MG 53909-1G 53909-5G
77335-764EA 365.65 USD
77335-764 77352-936 77377-160
1-(2-Methoxy-3,5-dimethylphenyl)ethanol ≥97%
1-(2-Methoxy-3,5-dimethylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34757175
CAS Number: 1367750-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR