1,2-Dichloro-4-iodo-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

53900-500MG 53900-1G 53900-5G
77329-132EA 310.03 USD
77329-132 77352-934 77377-158
1,2-Dichloro-4-iodo-5-methylbenzene ≥95%
1,2-Dichloro-4-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂I
MW: 286.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786707
CAS Number: 1806312-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR