1-Iodo-2-methoxy-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

53894-500MG 53894-1G 53894-5G
77335-404EA 353.29 USD
77335-404 77352-932 77377-156
1-Iodo-2-methoxy-3,5-dimethylbenzene ≥97%
1-Iodo-2-methoxy-3,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
MW: 262.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743671
CAS Number: 65434-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR