2-Hydroxy-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53886-500MG 53886-1G 53886-5G 53886-25G
77329-130EA 184.37 USD
77329-130 77347-160 77365-424 77387-432
2-Hydroxy-3,5-dimethylbenzoic acid ≥97%
2-Hydroxy-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00667775
CAS Number: 6245-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR