(2,6-Dimethyl-4-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53877-500MG 53877-1G 53877-5G
77337-514EA 416.12 USD
77337-514 77352-930 77377-154
(2,6-Dimethyl-4-(methylthio)phenyl)methanol ≥97%
(2,6-Dimethyl-4-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD34564245
CAS Number: 2226776-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR