(4-Iodo-2,6-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53875-500MG 53875-1G 53875-5G 53875-25G
77329-128EA 147.29 USD
77329-128 77345-210 77365-422 77387-430
(4-Iodo-2,6-dimethylphenyl)methanol ≥97%
(4-Iodo-2,6-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD34564244
CAS Number: 2383994-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR