2-Isopropoxy-4-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53860-500MG 53860-1G 53860-5G 53860-25G
77329-126EA 126.69 USD
77329-126 77344-132 77365-420 77377-152
2-Isopropoxy-4-(trifluoromethoxy)benzaldehyde ≥97%
2-Isopropoxy-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₃
MW: 248.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757165
CAS Number: 1071170-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR