(3-Bromo-5-chloro-2,6-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53855-500MG 53855-1G 53855-5G 53855-25G
77329-124EA 96.82 USD
77329-124 77342-644 77365-418 77377-150
(3-Bromo-5-chloro-2,6-difluorophenyl)(methyl)sulfane ≥97%
(3-Bromo-5-chloro-2,6-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₄BrClF₂S
MW: 273.525 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757163
CAS Number: 2755723-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR