5-Fluoro-4-iodo-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53824-500MG 53824-1G 53824-5G 53824-25G
77329-118EA 162.74 USD
77329-118 77346-088 77365-414 77387-424
5-Fluoro-4-iodo-2-methylbenzaldehyde ≥97%
5-Fluoro-4-iodo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD34757158
CAS Number: 2383912-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR