4'-Bromo-2-methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

53818-500MG 53818-1G 53818-5G 53818-25G
77329-116EA 180.25 USD
77329-116 77346-754 77365-412 77387-422
4'-Bromo-2-methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
4'-Bromo-2-methoxy-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O
MW: 331.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757153
CAS Number: 2764731-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR