Methyl 2,3,4-trichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

53817-500MG 53817-1G 53817-5G 53817-25G
77329-114EA 281.19 USD
77329-114 77352-922 77365-410 77387-420
Methyl 2,3,4-trichlorobenzoate ≥97%
Methyl 2,3,4-trichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Cl₃O₂
MW: 239.483 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399689
CAS Number: 89978-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR